Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGGGACCCTGTGCTGCATGCC[A/G]ACAAGGCGCGTGGCCAGCAGAACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607837 | ||||||||||||||||||||
Literature Links: |
CLN8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLN8 - ceroid-lipofuscinosis, neuronal 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018941.3 | 529 | Missense Mutation | AAC,GAC | N,D 94 | NP_061764.2 | |
XM_005266021.3 | 529 | Missense Mutation | AAC,GAC | N,D 94 | XP_005266078.1 | |
XM_005266022.1 | 529 | Missense Mutation | AAC,GAC | N,D 94 | XP_005266079.1 | |
XM_005266023.1 | 529 | Missense Mutation | AAC,GAC | N,D 94 | XP_005266080.1 | |
XM_011534745.1 | 529 | Missense Mutation | AAC,GAC | N,D 94 | XP_011533047.1 | |
XM_011534746.2 | 529 | Missense Mutation | AAC,GAC | N,D 94 | XP_011533048.1 | |
XM_011534747.1 | 529 | Missense Mutation | AAC,GAC | N,D 94 | XP_011533049.1 |
LOC101927752 - uncharacterized LOC101927752 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |