Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGACTCCTGACCTCCACAGCCA[G/A]CATGATGTAGGCAATGATCAACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607338 MIM: 611940 | ||||||||||||||||||||
Literature Links: |
LEPROTL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LEPROTL1 - leptin receptor overlapping transcript-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128208.1 | 1266 | Intron | NP_001121680.1 | |||
NM_015344.2 | 1266 | Intron | NP_056159.2 |
MBOAT4 - membrane bound O-acyltransferase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100916.1 | 1266 | Silent Mutation | CTG,TTG | L,L 395 | NP_001094386.1 |