Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCTGCCCTGGACGCTGGCAGCA[A/G]TGACAGCAGCAGCCGCCAGCACCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603398 | ||||||||||||||||||||
Literature Links: |
WISP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WISP1 - WNT1 inducible signaling pathway protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204869.1 | 140 | Missense Mutation | ATG,GTG | M,V 12 | NP_001191798.1 | |
NM_001204870.1 | 140 | Missense Mutation | ATG,GTG | M,V 12 | NP_001191799.1 | |
NM_003882.3 | 140 | Missense Mutation | ATG,GTG | M,V 12 | NP_003873.1 | |
NM_080838.2 | 140 | Missense Mutation | ATG,GTG | M,V 12 | NP_543028.1 |