Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGAAAAGAAAAGGGATCTGGGC[A/G]TAGCAGAATCAGGAGACCTTTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604782 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASH2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASH2L - ASH2 like histone lysine methyltransferase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105214.2 | Intron | NP_001098684.1 | ||||
NM_001261832.1 | Intron | NP_001248761.1 | ||||
NM_001282272.1 | Intron | NP_001269201.1 | ||||
NM_004674.4 | Intron | NP_004665.2 | ||||
XM_005273682.1 | Intron | XP_005273739.1 | ||||
XM_005273683.1 | Intron | XP_005273740.1 | ||||
XM_006716412.1 | Intron | XP_006716475.1 | ||||
XM_006716413.2 | Intron | XP_006716476.1 |