Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGACTTTCCTTGGGAAATCATG[A/G]GTCTTTTATTTTTGCAATTTGGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602232 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNQ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNQ3 - potassium voltage-gated channel subfamily Q member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204824.1 | 10633 | UTR 3 | NP_001191753.1 | |||
NM_004519.3 | 10633 | UTR 3 | NP_004510.1 | |||
XM_005250914.3 | 10633 | UTR 3 | XP_005250971.1 | |||
XM_006716555.3 | 10633 | UTR 3 | XP_006716618.1 | |||
XM_011517026.2 | 10633 | UTR 3 | XP_011515328.1 | |||
XM_017013400.1 | 10633 | UTR 3 | XP_016868889.1 |