Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTCGCTGAGGAAAAGAAGATAC[A/G]TCATTCTTCTAAGAGAACCACATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC45A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC45A4 - solute carrier family 45 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080431.2 | 2836 | Missense Mutation | ACG,ATG | T,M 759 | NP_001073900.1 | |
NM_001286646.1 | 2836 | UTR 3 | NP_001273575.1 | |||
NM_001286648.1 | 2836 | Intron | NP_001273577.1 | |||
XM_011517169.2 | 2836 | UTR 3 | XP_011515471.1 | |||
XM_011517170.2 | 2836 | UTR 3 | XP_011515472.1 | |||
XM_011517171.2 | 2836 | UTR 3 | XP_011515473.1 | |||
XM_011517172.2 | 2836 | UTR 3 | XP_011515474.1 | |||
XM_011517173.2 | 2836 | Intron | XP_011515475.1 | |||
XM_011517174.2 | 2836 | UTR 3 | XP_011515476.1 | |||
XM_011517176.2 | 2836 | UTR 3 | XP_011515478.1 | |||
XM_011517177.2 | 2836 | UTR 3 | XP_011515479.1 | |||
XM_011517178.2 | 2836 | UTR 3 | XP_011515480.1 | |||
XM_011517179.2 | 2836 | UTR 3 | XP_011515481.1 | |||
XM_017013648.1 | 2836 | UTR 3 | XP_016869137.1 | |||
XM_017013649.1 | 2836 | UTR 3 | XP_016869138.1 | |||
XM_017013650.1 | 2836 | UTR 3 | XP_016869139.1 |