Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAAGATGCACTTTGACTTCCTG[A/T]TAGAGTCGTGTCAGGTGGGAATGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 132811 | ||||||||||||||||||||
Literature Links: |
EPHX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPHX2 - epoxide hydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256482.1 | 490 | Missense Mutation | ATA,TTA | I,L 98 | NP_001243411.1 | |
NM_001256483.1 | 490 | Missense Mutation | ATA,TTA | I,L 85 | NP_001243412.1 | |
NM_001256484.1 | 490 | Missense Mutation | ATA,TTA | I,L 98 | NP_001243413.1 | |
NM_001979.5 | 490 | Missense Mutation | ATA,TTA | I,L 151 | NP_001970.2 | |
XM_017013199.1 | 490 | Missense Mutation | ATA,TTA | I,L 151 | XP_016868688.1 | |
XM_017013200.1 | 490 | Missense Mutation | ATA,TTA | I,L 151 | XP_016868689.1 |