Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCACAGTTTTGCCCCAAATGTC[A/G]TACTGTGGTTTCTTAGCGCCGATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 103070 | ||||||||||||||||||||
Literature Links: |
ADCY8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADCY8 - adenylate cyclase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115.2 | 3709 | Silent Mutation | TAC,TAT | Y,Y 1108 | NP_001106.1 | |
XM_005250769.3 | 3709 | Silent Mutation | TAC,TAT | Y,Y 1078 | XP_005250826.1 | |
XM_006716501.3 | 3709 | Silent Mutation | TAC,TAT | Y,Y 1042 | XP_006716564.1 | |
XM_017013006.1 | 3709 | Silent Mutation | TAC,TAT | Y,Y 1012 | XP_016868495.1 | |
XM_017013007.1 | 3709 | Intron | XP_016868496.1 |