Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGAGGCGGTATTGACCGAAGAG[C/G]TTGATGAGGAAGAGCAGCTGCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612021 | ||||||||||||||||||||
Literature Links: |
OTUD6B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OTUD6B - OTU domain containing 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286745.1 | 105 | UTR 5 | NP_001273674.1 | |||
NM_016023.3 | 105 | Missense Mutation | CTT,GTT | L,V 39 | NP_057107.3 | |
XM_011517129.2 | 105 | UTR 5 | XP_011515431.2 |
OTUD6B-AS1 - OTUD6B antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |