Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGCACTTGAGGAACATTGTGTC[A/G]TCCTTCACGTAGGCGTGCTTGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609740 MIM: 601711 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHF19 - PHD finger protein 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAF1 - TNF receptor associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190945.1 | 1634 | Silent Mutation | NP_001177874.1 | |||
NM_001190947.1 | 1634 | Silent Mutation | NP_001177876.1 | |||
NM_005658.4 | 1634 | Silent Mutation | NP_005649.1 |