Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGGGGAGCTGGAGGGACAGCCG[A/G]CGCTGGATGCTGAAGAGAAGGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607340 | ||||||||||||||||||||
Literature Links: |
GABBR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GABBR2 - gamma-aminobutyric acid type B receptor subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005458.7 | 2548 | Silent Mutation | CGC,CGT | R,R 890 | NP_005449.5 | |
XM_005252316.4 | 2548 | Silent Mutation | CGC,CGT | R,R 632 | XP_005252373.1 | |
XM_017015331.1 | 2548 | Silent Mutation | CGC,CGT | R,R 792 | XP_016870820.1 | |
XM_017015332.1 | 2548 | Silent Mutation | CGC,CGT | R,R 632 | XP_016870821.1 |