Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGTCCTCAAACAGGCCGTGGTC[A/G]GTCTCAGGGTAGCCTGGGCAGAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601309 | ||||||||||||||||||||
Literature Links: |
PTCH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTCH1 - patched 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000264.3 | 4200 | Silent Mutation | ACC,ACT | T,T 1404 | NP_000255.2 | |
NM_001083602.1 | 4200 | Silent Mutation | ACC,ACT | T,T 1338 | NP_001077071.1 | |
NM_001083603.1 | 4200 | Silent Mutation | ACC,ACT | T,T 1403 | NP_001077072.1 | |
NM_001083604.1 | 4200 | Silent Mutation | ACC,ACT | T,T 1253 | NP_001077073.1 | |
NM_001083605.1 | 4200 | Silent Mutation | ACC,ACT | T,T 1253 | NP_001077074.1 | |
NM_001083606.1 | 4200 | Silent Mutation | ACC,ACT | T,T 1253 | NP_001077075.1 | |
NM_001083607.1 | 4200 | Silent Mutation | ACC,ACT | T,T 1253 | NP_001077076.1 |