Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAGCAGTGCTGCCAGCCACTGC[A/G]CACCTGCCCGCTGTGCCGCCAGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614045 MIM: 610933 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM129B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM129B - family with sequence similarity 129 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRSAM1 - leucine rich repeat and sterile alpha motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005373.3 | 3482 | Missense Mutation | CAC,CGC | H,R 704 | NP_001005373.1 | |
NM_001005374.3 | 3482 | Missense Mutation | CAC,CGC | H,R 704 | NP_001005374.1 | |
NM_001190723.2 | 3482 | Missense Mutation | CAC,CGC | H,R 677 | NP_001177652.1 | |
NM_138361.5 | 3482 | Missense Mutation | CAC,CGC | H,R 704 | NP_612370.3 | |
XM_006717316.3 | 3482 | Missense Mutation | CAC,CGC | H,R 671 | XP_006717379.1 | |
XM_017015283.1 | 3482 | Missense Mutation | CAC,CGC | H,R 704 | XP_016870772.1 | |
XM_017015284.1 | 3482 | Missense Mutation | CAC,CGC | H,R 441 | XP_016870773.1 |