Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCTTCAAATCAGAATCTTCCAA[C/T]ACTTTCTGGACGGGCTTCATAGTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138120 MIM: 605962 | ||||||||||||||||||||
Literature Links: |
HSPA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSPA5 - heat shock protein family A (Hsp70) member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005347.4 | 1296 | Silent Mutation | NP_005338.1 |
RABEPK - Rab9 effector protein with kelch motifs | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |