Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCAAAGATAGTTTGGAGAGCAC[A/G]GACCACATGTTGGGTAGGAAACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606443 MIM: 609471 MIM: 186745 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CREB3 - cAMP responsive element binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006368.4 | 2841 | Intron | NP_006359.3 |
GBA2 - glucosylceramidase beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020944.2 | 2841 | Missense Mutation | CGT,TGT | R,C 780 | NP_065995.1 | |
XM_005251526.4 | 2841 | Missense Mutation | CGT,TGT | R,C 764 | XP_005251583.1 | |
XM_006716809.3 | 2841 | Missense Mutation | CGT,TGT | R,C 786 | XP_006716872.1 | |
XM_011517973.2 | 2841 | Missense Mutation | CGT,TGT | R,C 780 | XP_011516275.1 | |
XM_017014937.1 | 2841 | Missense Mutation | CGT,TGT | R,C 758 | XP_016870426.1 | |
XM_017014938.1 | 2841 | Missense Mutation | CGT,TGT | R,C 786 | XP_016870427.1 | |
XM_017014939.1 | 2841 | Missense Mutation | CGT,TGT | R,C 758 | XP_016870428.1 | |
XM_017014940.1 | 2841 | Missense Mutation | CGT,TGT | R,C 707 | XP_016870429.1 | |
XM_017014941.1 | 2841 | Missense Mutation | CGT,TGT | R,C 707 | XP_016870430.1 | |
XM_017014942.1 | 2841 | Missense Mutation | CGT,TGT | R,C 635 | XP_016870431.1 | |
XM_017014943.1 | 2841 | Missense Mutation | CGT,TGT | R,C 629 | XP_016870432.1 | |
XM_017014944.1 | 2841 | Missense Mutation | CGT,TGT | R,C 601 | XP_016870433.1 | |
XM_017014945.1 | 2841 | Missense Mutation | CGT,TGT | R,C 595 | XP_016870434.1 | |
XM_017014946.1 | 2841 | Missense Mutation | CGT,TGT | R,C 493 | XP_016870435.1 |
MIR6853 - microRNA 6853 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLN1 - talin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |