Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACAAGATCCGGCCCATCTCCAGC[A/G]TTGGACAGGTGGGGCTCCCGACCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615365 MIM: 605284 | ||||||||||||||||||||
Literature Links: |
AK8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AK8 - adenylate kinase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPACA9 - sperm acrosome associated 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316897.1 | 297 | Missense Mutation | ATT,GTT | I,V 46 | NP_001303826.1 | |
NM_001316898.1 | 297 | Missense Mutation | ATT,GTT | I,V 46 | NP_001303827.1 | |
NM_001316900.1 | 297 | Missense Mutation | ATT,GTT | I,V 46 | NP_001303829.1 | |
NM_018956.4 | 297 | Missense Mutation | ATT,GTT | I,V 46 | NP_061829.3 | |
XM_017014231.1 | 297 | Missense Mutation | ATT,GTT | I,V 46 | XP_016869720.1 |
TSC1 - tuberous sclerosis 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |