Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGCTAGAGGTTCTCCTCCCAAT[A/G]AAGCCTCTGAGTACCTAACACTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608040 MIM: 603873 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT74 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFT74 - intraflagellar transport 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099222.1 | Intron | NP_001092692.1 | ||||
NM_001099223.1 | Intron | NP_001092693.1 | ||||
NM_001099224.1 | Intron | NP_001092694.1 | ||||
NM_025103.2 | Intron | NP_079379.2 | ||||
XM_011518035.2 | Intron | XP_011516337.1 | ||||
XM_011518036.2 | Intron | XP_011516338.1 | ||||
XM_017015163.1 | Intron | XP_016870652.1 | ||||
XM_017015164.1 | Intron | XP_016870653.1 |
IFT74-AS1 - IFT74 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLAA - phospholipase A2 activating protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |