Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGTGGAGGAGCTGCTTCGTGG[A/G]GCTCAGTATTTCAACACACCACGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603960 MIM: 607141 | ||||||||||||||||||||
Literature Links: |
CCIN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCIN - calicin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005893.2 | 432 | Silent Mutation | GGA,GGG | G,G 107 | NP_005884.2 |
GLIPR2 - GLI pathogenesis related 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |