Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCTTAACAATCTCTTCTACTT[A/C]TGCCTCCACCGTAACCTTTGTACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605046 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBQLN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBQLN1 - ubiquilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013438.4 | 3903 | UTR 3 | NP_038466.2 | |||
NM_053067.2 | 3903 | UTR 3 | NP_444295.1 | |||
XM_005251948.2 | 3903 | Intron | XP_005252005.1 |