Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACCAGTCGCGGGCCTTTGGGTG[A/C]GTGAAGTCTAGCACCGCGCCGATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf24 - chromosome 9 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA1161 - KIAA1161 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020702.4 | 1475 | Silent Mutation | ACG,ACT | T,T 436 | NP_065753.2 | |
XM_011517966.2 | 1475 | Silent Mutation | ACG,ACT | T,T 436 | XP_011516268.1 | |
XM_017014930.1 | 1475 | Silent Mutation | ACG,ACT | T,T 436 | XP_016870419.1 |