Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATCTCTGCTGCTCCCACTGGAG[A/C]GTGAGTCTGGAGTCCCTGGCACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614730 MIM: 608292 | ||||||||||||||||||||
Literature Links: |
FAM214B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM214B - family with sequence similarity 214 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGO - phosphatidylinositol glycan anchor biosynthesis class O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STOML2 - stomatin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287031.1 | 982 | Missense Mutation | CGC,CTC | R,L 286 | NP_001273960.1 | |
NM_001287032.1 | 982 | Missense Mutation | CGC,CTC | R,L 280 | NP_001273961.1 | |
NM_001287033.1 | 982 | Missense Mutation | CGC,CTC | R,L 285 | NP_001273962.1 | |
NM_013442.2 | 982 | Missense Mutation | CGC,CTC | R,L 331 | NP_038470.1 |