Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGCTTGACTTCTATTGTATTCC[C/T]GCAATTAAGAGAACTCCCCTACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMD3 - FERM domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244959.1 | Intron | NP_001231888.1 | ||||
NM_001244960.1 | Intron | NP_001231889.1 | ||||
NM_001244961.1 | Intron | NP_001231890.1 | ||||
NM_001244962.1 | Intron | NP_001231891.1 | ||||
NM_174938.5 | Intron | NP_777598.3 | ||||
XM_017014588.1 | Intron | XP_016870077.1 | ||||
XM_017014589.1 | Intron | XP_016870078.1 | ||||
XM_017014590.1 | Intron | XP_016870079.1 | ||||
XM_017014591.1 | Intron | XP_016870080.1 |
LOC102723989 - uncharacterized LOC102723989 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |