Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGGACCAGCAGTCAGCAGGAA[A/G]GACGTCCCATCGTCCACATCAGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608167 MIM: 610224 | ||||||||||||||||||||
Literature Links: |
KCNT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNT1 - potassium sodium-activated channel subfamily T member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOHLH1 - spermatogenesis and oogenesis specific basic helix-loop-helix 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012415.2 | 2049 | Silent Mutation | TCC,TCT | S,S 304 | NP_001012415.2 | |
NM_001101677.1 | 2049 | Silent Mutation | TCC,TCT | S,S 304 | NP_001095147.1 | |
XM_005263403.3 | 2049 | Silent Mutation | TCC,TCT | S,S 304 | XP_005263460.1 | |
XM_006717109.3 | 2049 | Silent Mutation | TCC,TCT | S,S 236 | XP_006717172.1 | |
XM_011518698.2 | 2049 | Silent Mutation | TCC,TCT | S,S 304 | XP_011517000.1 |