Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGGGCGCACCCCGATGCCGAGCC[C/T]GCAGCTTCTGGTGCTCTTCGGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606073 MIM: 616499 MIM: 613354 | ||||||||||||||||||||
Literature Links: |
NDOR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDOR1 - NADPH dependent diflavin oxidoreductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144026.2 | 101 | Missense Mutation | CCG,CTG | P,L 4 | NP_001137498.1 | |
NM_001144027.2 | 101 | Missense Mutation | CCG,CTG | P,L 4 | NP_001137499.1 | |
NM_001144028.2 | 101 | Missense Mutation | CCG,CTG | P,L 4 | NP_001137500.1 | |
NM_014434.3 | 101 | Missense Mutation | CCG,CTG | P,L 4 | NP_055249.1 | |
XM_011518545.1 | 101 | Missense Mutation | CCG,CTG | P,L 4 | XP_011516847.1 | |
XM_011518547.1 | 101 | Intron | XP_011516849.1 | |||
XM_011518548.2 | 101 | UTR 5 | XP_011516850.1 | |||
XM_017014626.1 | 101 | Missense Mutation | CCG,CTG | P,L 4 | XP_016870115.1 | |
XM_017014627.1 | 101 | Missense Mutation | CCG,CTG | P,L 4 | XP_016870116.1 |
TMEM203 - transmembrane protein 203 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053045.1 | 101 | Intron | NP_444273.1 |
TPRN - taperin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |