Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCAGAGGAGGACTGCATGCAG[C/G]TGAACCCCTCCTTCAAGGGCATCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM78A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM78A - family with sequence similarity 78 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLPP7 - phospholipid phosphatase 7 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032728.3 | 533 | Missense Mutation | CTG,GTG | L,V 73 | NP_116117.3 | |
XM_005272230.3 | 533 | Missense Mutation | CTG,GTG | L,V 73 | XP_005272287.1 | |
XM_005272231.1 | 533 | Missense Mutation | CTG,GTG | L,V 73 | XP_005272288.1 |