Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGTAGGAATGGTGGGGGTGCA[C/T]AGCCAGGGGCCGCTCCCGAGGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612506 | ||||||||||||||||||||
Literature Links: |
UBAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBAP2 - ubiquitin associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282529.1 | 3289 | Missense Mutation | TAT,TGT | Y,C 792 | NP_001269458.1 | |
NM_001282530.1 | 3289 | Missense Mutation | TAT,TGT | Y,C 298 | NP_001269459.1 | |
NM_018449.3 | 3289 | Missense Mutation | TAT,TGT | Y,C 1059 | NP_060919.3 |
UBE2R2 - ubiquitin conjugating enzyme E2 R2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |