Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCAGACTGCGTGTGCAACGTG[C/G]AGTATTTCACTAGGGGAAAAAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300846 MIM: 300171 | ||||||||||||||||||||
Literature Links: |
CD99L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD99L2 - CD99 molecule like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184808.1 | 812 | Missense Mutation | TCC,TGC | S,C 171 | NP_001171737.1 | |
NM_001242614.1 | 812 | Missense Mutation | TCC,TGC | S,C 254 | NP_001229543.1 | |
NM_031462.3 | 812 | Missense Mutation | TCC,TGC | S,C 244 | NP_113650.2 | |
NM_134445.3 | 812 | Missense Mutation | TCC,TGC | S,C 172 | NP_604394.1 | |
NM_134446.3 | 812 | Missense Mutation | TCC,TGC | S,C 195 | NP_604395.1 | |
XM_011531202.2 | 812 | Intron | XP_011529504.1 | |||
XM_017029890.1 | 812 | Intron | XP_016885379.1 | |||
XM_017029891.1 | 812 | Intron | XP_016885380.1 |
MTMR1 - myotubularin related protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |