Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGTTGTTGACAAAACTAAGAAA[G/T]TCAAGGTGTGCAGGCACTGAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300457 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NHS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NHS - NHS actin remodeling regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136024.3 | Intron | NP_001129496.1 | ||||
NM_001291867.1 | Intron | NP_001278796.1 | ||||
NM_001291868.1 | Intron | NP_001278797.1 | ||||
NM_198270.3 | Intron | NP_938011.1 | ||||
XM_011545528.2 | Intron | XP_011543830.1 |