Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGTCTTTCTGTAAACCATTTCT[C/T]GAAAACTGGCCAGAATCTTGTTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300462 MIM: 300766 | ||||||||||||||||||||
Literature Links: |
AKAP14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKAP14 - A-kinase anchoring protein 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKAP - NFKB activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024528.3 | 1372 | Missense Mutation | CAA,CGA | Q,R 402 | NP_078804.2 | |
XM_017029842.1 | 1372 | Missense Mutation | CAA,CGA | Q,R 303 | XP_016885331.1 |