Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTGGGCCAGGAAGCCAAGAGC[C/T]GTTGCTGAAAAGATGGTGGTCAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 309860 | ||||||||||||||||||||
Literature Links: |
MAOB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAOB - monoamine oxidase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000898.4 | 1651 | Silent Mutation | ACA,ACG | T,T 505 | NP_000889.3 | |
XM_005272608.3 | 1651 | Silent Mutation | ACA,ACG | T,T 489 | XP_005272665.1 | |
XM_017029523.1 | 1651 | Silent Mutation | ACA,ACG | T,T 489 | XP_016885012.1 | |
XM_017029524.1 | 1651 | Silent Mutation | ACA,ACG | T,T 489 | XP_016885013.1 |