Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGATCTGATCCTGGACCACGCA[A/G]GGGGAAACAGAGCCTCCAGGGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300656 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UPRT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UPRT - uracil phosphoribosyltransferase homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307944.1 | 314 | Missense Mutation | AGG,GGG | R,G 40 | NP_001294873.1 | |
NM_145052.3 | 314 | Missense Mutation | AGG,GGG | R,G 40 | NP_659489.1 | |
XM_011530867.2 | 314 | Missense Mutation | AGG,GGG | R,G 40 | XP_011529169.1 |