Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGACCTGCTGGCCGTGGTGGC[C/T]AGCCCGGGCTCCGGGGCCTGGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM155B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM155B - family with sequence similarity 155 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015686.2 | 747 | Silent Mutation | GCC,GCT | A,A 233 | NP_056501.2 | |
XM_011530908.2 | 747 | Silent Mutation | GCC,GCT | A,A 233 | XP_011529210.1 | |
XM_011530909.2 | 747 | Silent Mutation | GCC,GCT | A,A 233 | XP_011529211.1 |