Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACAGCAGGCCGAGGATGAGGGG[C/G]TGGCTGCGGCTGCAGAGACTGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300383 MIM: 311040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFP - complement factor properdin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELK1 - ELK1, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114123.2 | 730 | Missense Mutation | CCC,GCC | P,A 172 | NP_001107595.1 | |
NM_001257168.1 | 730 | Intron | NP_001244097.1 | |||
NM_005229.4 | 730 | Missense Mutation | CCC,GCC | P,A 172 | NP_005220.2 | |
XM_017029339.1 | 730 | Missense Mutation | CCC,GCC | P,A 172 | XP_016884828.1 |