Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACCCCAACCACCCTAGTTCCTC[C/T]GGTAAGAGTCTCTGACCCTGTGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 313470 MIM: 450000 | ||||||||||||||||||||
Literature Links: |
CD99 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD99 - CD99 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122898.2 | 489 | Silent Mutation | TCC,TCT | S,S 71 | NP_001116370.1 | |
NM_001321367.1 | 489 | Silent Mutation | TCC,TCT | S,S 87 | NP_001308296.1 | |
NM_001321368.1 | 489 | Silent Mutation | TCC,TCT | S,S 87 | NP_001308297.1 | |
NM_001321369.1 | 489 | Silent Mutation | TCC,TCT | S,S 71 | NP_001308298.1 | |
NM_001321370.1 | 489 | Silent Mutation | TCC,TCT | S,S 87 | NP_001308299.1 | |
NM_002414.4 | 489 | Silent Mutation | TCC,TCT | S,S 87 | NP_002405.1 |