Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTTATAGGGCTTCTCTCCAGTA[C/T]GCGTTCTCTGATGTATAATGAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300819 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF630 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF630 - zinc finger protein 630 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037735.3 | 1837 | Missense Mutation | CAT,CGT | H,R 425 | NP_001032824.2 | |
NM_001190255.2 | 1837 | Missense Mutation | CAT,CGT | H,R 411 | NP_001177184.1 | |
NM_001282201.1 | 1837 | Missense Mutation | CAT,CGT | H,R 425 | NP_001269130.1 | |
NM_001282202.1 | 1837 | Missense Mutation | CAT,CGT | H,R 301 | NP_001269131.1 |
ZNF630-AS1 - ZNF630 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |