Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACGGTACTCTGGCTGAGGTACA[C/T]GATTCAGGACAGACTCCACATGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300269 MIM: 311870 | ||||||||||||||||||||
Literature Links: |
HDAC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HDAC8 - histone deacetylase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHKA1 - phosphorylase kinase regulatory subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122670.1 | 3316 | Missense Mutation | CAT,CGT | H,R 1104 | NP_001116142.1 | |
NM_001172436.1 | 3316 | Missense Mutation | CAT,CGT | H,R 1045 | NP_001165907.1 | |
NM_002637.3 | 3316 | Missense Mutation | CAT,CGT | H,R 1117 | NP_002628.2 | |
XM_006724661.2 | 3316 | Missense Mutation | CAT,CGT | H,R 1058 | XP_006724724.1 |