Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACAAATCCAGAATCCAAGCAAAT[A/C]GTAGAAGAGAAAACTAAAAGACAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300768 | ||||||||||||||||||||
Literature Links: |
CYLC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYLC1 - cylicin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271680.1 | 495 | Intron | NP_001258609.1 | |||
NM_021118.2 | 495 | Silent Mutation | ATA,ATC | I,I 147 | NP_066941.1 | |
XM_005262086.3 | 495 | Silent Mutation | ATA,ATC | I,I 146 | XP_005262143.1 |