Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCTTGATCTTCTCCATCTGCAG[A/G]GTCTTGTTCCTGCAATTTGCAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300897 | ||||||||||||||||||||
Literature Links: |
ZC4H2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZC4H2 - zinc finger C4H2-type containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178032.2 | 219 | UTR 5 | NP_001171503.1 | |||
NM_001178033.2 | 219 | Silent Mutation | ACC,ACT | T,T 21 | NP_001171504.1 | |
NM_001243804.1 | 219 | UTR 5 | NP_001230733.1 | |||
NM_018684.3 | 219 | Silent Mutation | ACC,ACT | T,T 21 | NP_061154.1 |