Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTCAGGCCTTCCTGTCAGGTGT[C/T]TGTCTCCCAGGTTCCTGTGAATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXorf38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXorf38 - chromosome X open reading frame 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144970.2 | 986 | Silent Mutation | NP_659407.1 | |||
XM_005272589.2 | 986 | Missense Mutation | XP_005272646.2 | |||
XM_006724527.3 | 986 | Silent Mutation | XP_006724590.2 | |||
XM_006724528.2 | 986 | Silent Mutation | XP_006724591.1 | |||
XM_017029302.1 | 986 | Silent Mutation | XP_016884791.1 | |||
XM_017029303.1 | 986 | Silent Mutation | XP_016884792.1 | |||
XM_017029304.1 | 986 | Silent Mutation | XP_016884793.1 | |||
XM_017029305.1 | 986 | Silent Mutation | XP_016884794.1 |
MPC1L - mitochondrial pyruvate carrier 1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |