Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAGAAGCTCTGCATTCTTTGGC[A/G]TTTTCAGTAGAAAACACTGAAACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 306480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PUDP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PUDP - pseudouridine 5'-phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135565.1 | Intron | NP_001129037.1 | ||||
NM_001178135.1 | Intron | NP_001171606.1 | ||||
NM_001178136.1 | Intron | NP_001171607.1 | ||||
NM_012080.4 | Intron | NP_036212.3 |