Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTTACTCTTAATCTCTTCCAGG[C/G]TGTGCTGGATCCCATAGCCGAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300443 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC7A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC7A3 - solute carrier family 7 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048164.2 | 1940 | Missense Mutation | ACC,AGC | T,S 589 | NP_001041629.1 | |
NM_032803.5 | 1940 | Missense Mutation | ACC,AGC | T,S 589 | NP_116192.4 | |
XM_017029912.1 | 1940 | Missense Mutation | ACC,AGC | T,S 589 | XP_016885401.1 |