Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAGGAATGTGGGGGTGGCATCA[C/T]CTAAATTTTAATAGGTTGGAACTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300191 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN6 - tetraspanin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278740.1 | Intron | NP_001265669.1 | ||||
NM_001278741.1 | Intron | NP_001265670.1 | ||||
NM_001278742.1 | Intron | NP_001265671.1 | ||||
NM_001278743.1 | Intron | NP_001265672.1 | ||||
NM_003270.3 | Intron | NP_003261.1 | ||||
XM_011531018.1 | Intron | XP_011529320.1 |