Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCATCGCCGGCCTCGTGGTGTTC[C/G]TCACGGCCAACGTGCTGCGGGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300371 MIM: 300398 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCD1 - ATP binding cassette subfamily D member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000033.3 | 1156 | Missense Mutation | CTC,GTC | L,V 253 | NP_000024.2 |
BCAP31 - B-cell receptor-associated protein 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |