Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGAGGAGCCCTTCCCACTCCTGG[C/G]ACCTCCTCGCGGGGTGGGCACCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300941 | ||||||||||||||||||||
Literature Links: |
FAM199X PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM199X - family with sequence similarity 199, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207318.3 | 382 | Missense Mutation | GCA,GGA | A,G 25 | NP_997201.1 | |
XM_005262079.2 | 382 | Missense Mutation | GCA,GGA | A,G 25 | XP_005262136.1 | |
XM_017029270.1 | 382 | Intron | XP_016884759.1 |
SLC25A53 - solute carrier family 25 member 53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |