Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGAGTTGCTGACAGTGATGGC[C/T]GGTGTGGCCACGCCTTGACTACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 314310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TFE3 - transcription factor binding to IGHM enhancer 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282142.1 | 1233 | Silent Mutation | CCA,CCG | P,P 209 | NP_001269071.1 | |
NM_006521.5 | 1233 | Silent Mutation | CCA,CCG | P,P 314 | NP_006512.2 |