Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCGGCCCGGCGGGCCCGGGACC[A/T]GGAGCACCGCCCACAGCCGGCGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611171 MIM: 604294 | ||||||||||||||||||||
Literature Links: |
SHTN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHTN1 - shootin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAX1 - ventral anterior homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112704.1 | 998 | Silent Mutation | CCA,CCT | P,P 251 | NP_001106175.1 | |
NM_199131.2 | 998 | Intron | NP_954582.1 |