Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCATGGGGGGCGCTCAGCTGGA[A/G]CTACCGAGCGGTGCCAGGCCAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614587 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHAC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHAC1 - ChaC glutathione specific gamma-glutamylcyclotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142776.1 | 41 | Silent Mutation | GAA,GAG | E,E 7 | NP_001136248.1 | |
NM_024111.3 | 41 | Silent Mutation | GAA,GAG | E,E 7 | NP_077016.2 | |
XM_017022563.1 | 41 | Silent Mutation | GAA,GAG | E,E 7 | XP_016878052.1 | |
XM_017022564.1 | 41 | Silent Mutation | GAA,GAG | E,E 7 | XP_016878053.1 | |
XM_017022565.1 | 41 | Silent Mutation | GAA,GAG | E,E 7 | XP_016878054.1 |