Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCGGCCTCCTGGATGGCGACG[T/C]GGATCTGCACCGCCGTCTTGGGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611300 | ||||||||||||||||||||
Literature Links: |
LARP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LARP6 - La ribonucleoprotein domain family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286679.1 | 233 | Intron | NP_001273608.1 | |||
NM_018357.3 | 233 | Missense Mutation | CAC,CGC | H,R 19 | NP_060827.2 | |
NM_197958.2 | 233 | Missense Mutation | CAC,CGC | H,R 19 | NP_932062.1 |
LRRC49 - leucine rich repeat containing 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199017.2 | 233 | Intron | NP_001185946.1 | |||
NM_001199018.2 | 233 | Intron | NP_001185947.1 | |||
NM_001284357.1 | 233 | Intron | NP_001271286.1 | |||
NM_017691.4 | 233 | Intron | NP_060161.2 | |||
XM_005254492.4 | 233 | Intron | XP_005254549.1 | |||
XM_011521715.2 | 233 | Intron | XP_011520017.1 | |||
XM_011521717.2 | 233 | Intron | XP_011520019.1 | |||
XM_011521718.2 | 233 | Intron | XP_011520020.1 | |||
XM_017022357.1 | 233 | Intron | XP_016877846.1 | |||
XM_017022358.1 | 233 | Intron | XP_016877847.1 | |||
XM_017022359.1 | 233 | Intron | XP_016877848.1 |