Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGTATATACAGGAGGTGGAGGG[A/G]CAGCAGGAAGCAAGGAGCCTTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616666 | ||||||||||||||||||||
Literature Links: |
PHF12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHF12 - PHD finger protein 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEZ6 - seizure related 6 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098635.1 | 3162 | UTR 3 | NP_001092105.1 | |||
NM_001290202.1 | 3162 | UTR 3 | NP_001277131.1 | |||
NM_178860.4 | 3162 | UTR 3 | NP_849191.3 | |||
XM_011524315.1 | 3162 | Silent Mutation | TGC,TGT | C,C 984 | XP_011522617.1 | |
XM_011524317.2 | 3162 | UTR 3 | XP_011522619.1 |