Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACATTGATTGCCCTGGAAAATAT[G/A]TCAGTGAGGCTTTCCTACCACTTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107930 MIM: 615383 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DDC - dopa decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000790.3 | 1729 | UTR 3 | NP_000781.1 | |||
NM_001082971.1 | 1729 | UTR 3 | NP_001076440.1 | |||
NM_001242886.1 | 1729 | UTR 3 | NP_001229815.1 | |||
NM_001242887.1 | 1729 | UTR 3 | NP_001229816.1 | |||
NM_001242888.1 | 1729 | UTR 3 | NP_001229817.1 | |||
NM_001242889.1 | 1729 | UTR 3 | NP_001229818.1 | |||
NM_001242890.1 | 1729 | Intron | NP_001229819.1 | |||
XM_005271745.4 | 1729 | UTR 3 | XP_005271802.1 | |||
XM_011515161.2 | 1729 | UTR 3 | XP_011513463.2 |
FIGNL1 - fidgetin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |